4-Bromo-3-fluorobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7630-100G OS-7630-25G OS-7630-5G
COBBOS-7630-100GEA 0 GBP
COBBOS-7630-100G COBBOS-7630-25G COBBOS-7630-5G
4-Bromo-3-fluorobenzonitrile
4-Bromo-3-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD07782069
CAS Number: 133059-44-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR