5-Bromo-2-chloroanisole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0936-5G
OT-0936-25G
OT-0936-1G
COBBOT-0936-5GEA
0
GBP
COBBOT-0936-5G
COBBOT-0936-25G
COBBOT-0936-1G
5-Bromo-2-chloroanisole
5-Bromo-2-chloroanisole
Formula:
C₇H₆BrClO MW: 221.48 g/mol |
MDL Number:
MFCD00078659 CAS Number: 16817-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...