5-Bromo-2-chlorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7585-5G
OS-7585-25G
OS-7585-1G
OS-7585-100G
COBBOS-7585-5GEA
0
GBP
COBBOS-7585-5G
COBBOS-7585-25G
COBBOS-7585-1G
COBBOS-7585-100G
5-Bromo-2-chlorobenzonitrile
5-Bromo-2-chlorobenzonitrile
Formula:
C₇H₃BrClN MW: 216.46 g/mol |
MDL Number:
MFCD00672948 CAS Number: 57381-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...