3-Bromo-5-nitrotoluene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1052-1G
AN-1052-5G
COBBAN-1052-1GEA
0
GBP
COBBAN-1052-1G
COBBAN-1052-5G
3-Bromo-5-nitrotoluene
3-Bromo-5-nitrotoluene
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD06659815 CAS Number: 52488-28-5 EINECS: 257-955-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...