3-Bromobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Bromobenzonitrile

BK-4526-25G BK-4526-100G BK-4526-500G
COBBBK-4526-25GEA 0 GBP
COBBBK-4526-25G COBBBK-4526-100G COBBBK-4526-500G
3-Bromobenzonitrile
3-Bromobenzonitrile
Formula: C₇H₄BrN
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 37…40 °C
Density: 1.562 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR