3-Bromo-p-toluonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7601-5G OS-7601-25G OS-7601-100G
COBBOS-7601-5GEA 0 GBP
COBBOS-7601-5G COBBOS-7601-25G COBBOS-7601-100G
3-Bromo-p-toluonitrile
3-Bromo-p-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD06797818
CAS Number: 42872-74-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR