3-Bromo-5-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013214-25G 013214-5G 013214-10G
FLUO013214-25GEA 0 GBP
FLUO013214-25G FLUO013214-5G FLUO013214-10G
3-Bromo-5-fluorobenzonitrile
3-Bromo-5-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD04038227
CAS Number: 179898-34-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR