3-Bromo-2-hydroxybenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4594-1G
CA-4594-5G
COBBCA-4594-1GEA
0
GBP
COBBCA-4594-1G
COBBCA-4594-5G
3-Bromo-2-hydroxybenzonitrile
3-Bromo-2-hydroxybenzonitrile
Formula:
C₇H₄BrNO MW: 198.02 g/mol |
MDL Number:
MFCD11042854 CAS Number: 13073-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...