4-Bromo-2-chlorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4688-25G
CA-4688-5G
COBBCA-4688-25GEA
0
GBP
COBBCA-4688-25G
COBBCA-4688-5G
4-Bromo-2-chlorobenzonitrile
4-Bromo-2-chlorobenzonitrile
Formula:
C₇H₃BrClN MW: 216.46 g/mol Boiling Pt: 142…143 °C (10 mmHg) Melting Pt: 67…68 °C |
MDL Number:
MFCD00040883 CAS Number: 154607-01-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...