2-Bromo-4,5-difluorophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene

OS-7562-5G OS-7562-25G OS-7562-100G
COBBOS-7562-5GEA 0 GBP
COBBOS-7562-5G COBBOS-7562-25G COBBOS-7562-100G
2-Bromo-4,5-difluorophenol
2-Bromo-4,5-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
MDL Number: MFCD00070751
CAS Number: 166281-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR