2-Bromo-4-nitrotoluene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7461-25G
OS-7461-100G
COBBOS-7461-25GEA
0
GBP
COBBOS-7461-25G
COBBOS-7461-100G
2-Bromo-4-nitrotoluene
2-Bromo-4-nitrotoluene
Formula:
C₇H₆BrNO₂ Boiling Pt: 175 °C (38 mmHg) Melting Pt: 77,5 °C |
MDL Number:
MFCD00007195 CAS Number: 7745-93-9 EINECS: 231-809-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...