2-Bromo-3-fluorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-8094-5G
OT-8094-1G
COBBOT-8094-5GEA
0
GBP
COBBOT-8094-5G
COBBOT-8094-1G
2-Bromo-3-fluorobenzonitrile
2-Bromo-3-fluorobenzonitrile
Formula:
C₇H₃BrFN |
MDL Number:
MFCD07368773 CAS Number: 425379-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...