2-Bromo-6-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024190-1G 024190-5G
FLUO024190-1GEA 0 GBP
FLUO024190-1G FLUO024190-5G
2-Bromo-6-fluorobenzonitrile
2-Bromo-6-fluorobenzonitrile
Formula: C₇H₃BrFN
MDL Number: MFCD06797753
CAS Number: 79544-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR