3-Bromo-4-chlorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4746-1G
CA-4746-5G
COBBCA-4746-1GEA
0
GBP
COBBCA-4746-1G
COBBCA-4746-5G
3-Bromo-4-chlorobenzonitrile
3-Bromo-4-chlorobenzonitrile
Formula:
C₇H₃BrClN MW: 216.46 g/mol |
MDL Number:
MFCD09025661 CAS Number: 948549-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...