4-Bromo-3-chlorobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7575-5G OS-7575-1G
COBBOS-7575-5GEA 0 GBP
COBBOS-7575-5G COBBOS-7575-1G
4-Bromo-3-chlorobenzonitrile
4-Bromo-3-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD09834777
CAS Number: 57418-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR