3-Bromo-2,4-dichlorotoluene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0577-5G
OT-0577-1G
COBBOT-0577-5GEA
0
GBP
COBBOT-0577-5G
COBBOT-0577-1G
3-Bromo-2,4-dichlorotoluene
3-Bromo-2,4-dichlorotoluene
Formula:
C₇H₅BrCl₂ |
MDL Number:
MFCD00079712 CAS Number: 206559-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...