2-Nitro-p-toluidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Methyl-2-nitroaniline, 4-Amino-3-nitrotoluene, 2-Nitro-4-methylaniline
AN-1227-5G
AN-1227-100G
AN-1227-25G
COBBAN-1227-5GEA
0
GBP
COBBAN-1227-5G
COBBAN-1227-100G
COBBAN-1227-25G
2-Nitro-p-toluidine
2-Nitro-p-toluidine
Formula:
C₇H₈N₂O₂ Boiling Pt: 303 °C (1013 hPa) Melting Pt: 112…114 °C Density: 1.164 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007907 CAS Number: 89-62-3 EINECS: 201-924-9 UN: 2660 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...