4-Fluoro-3-(trifluoromethyl)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7462-100G OS-7462-25G
COBBOS-7462-100GEA 0 GBP
COBBOS-7462-100G COBBOS-7462-25G
4-Fluoro-3-(trifluoromethyl)aniline
4-Fluoro-3-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
Boiling Pt: 207 °C (1013 hPa)
Density: 1.39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007834
CAS Number: 2357-47-3
EINECS: 219-095-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR