4-Fluoro-2-iodoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7693-5G OS-7693-25G OS-7693-100G
COBBOS-7693-5GEA 0 GBP
COBBOS-7693-5G COBBOS-7693-25G COBBOS-7693-100G
4-Fluoro-2-iodoaniline
4-Fluoro-2-iodoaniline
Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR