2-Amino-2',5-dichlorobenzophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone
M-Clarity™ quality level = MQ100
105155-10G
105155-50G
SIAL105155-10GEA
72.9
GBP
SIAL105155-10G
SIAL105155-50G
2-Amino-2',5-dichlorobenzophenone, Sigma-Aldrich®
2-Amino-2',5-dichlorobenzophenone
Formula:
C₁₃H₉Cl₂NO Boiling Pt: 385 °C (1013 hPa) Melting Pt: 88…91 °C |
MDL Number:
MFCD00007840 CAS Number: 2958-36-3 EINECS: 220-985-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...