4-Bromo-2-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene
OT-0799-100G
OT-0799-25G
OT-0799-5G
COBBOT-0799-100GEA
0
GBP
COBBOT-0799-100G
COBBOT-0799-25G
COBBOT-0799-5G
4-Bromo-2-(trifluoromethyl)aniline
4-Bromo-2-(trifluoromethyl)aniline
Formula:
C₇H₅BrF₃N MW: 240.02 g/mol Boiling Pt: 60.7…62 °C (1 mmHg) Density: 1.712 g/cm³ (25 °C) |
MDL Number:
MFCD00064393 CAS Number: 445-02-3 EINECS: 207-150-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...