4,5-Dichloro-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
HC-6205-25G
HC-6205-100G
HC-6205-5G
COBBHC-6205-25GEA
0
GBP
COBBHC-6205-25G
COBBHC-6205-100G
COBBHC-6205-5G
4,5-Dichloro-2-nitroaniline
4,5-Dichloro-2-nitroaniline
Formula:
C₆H₄Cl₂N₂O₂ MW: 207.02 g/mol Melting Pt: 180 Storage Temperature: Ambient |
MDL Number:
MFCD00007770 CAS Number: 6641-64-1 EINECS: 229-657-3 UN: 2237 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...