2-Amino-3-nitrophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxy-6-nitroaniline

AN-1414-5G AN-1414-1G AN-1414-25G AN-1414-100G
COBBAN-1414-5GEA 0 GBP
COBBAN-1414-5G COBBAN-1414-1G COBBAN-1414-25G COBBAN-1414-100G
2-Amino-3-nitrophenol
2-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MDL Number: MFCD00010875
CAS Number: 603-85-0
EINECS: 210-060-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR