3-Amino-4-chlorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1543-25G
AN-1543-5G
AN-1543-1G
COBBAN-1543-25GEA
0
GBP
COBBAN-1543-25G
COBBAN-1543-5G
COBBAN-1543-1G
3-Amino-4-chlorobenzonitrile
3-Amino-4-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD00017089 CAS Number: 53312-79-1 EINECS: 258-471-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...