6-Bromo-m-toluidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7482-25G
OS-7482-100G
COBBOS-7482-25GEA
0
GBP
COBBOS-7482-25G
COBBOS-7482-100G
6-Bromo-m-toluidine
6-Bromo-m-toluidine
Formula:
C₇H₈BrN |
MDL Number:
MFCD01806278 CAS Number: 53078-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...