2-Bromo-4-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1442-5G AN-1442-1G
COBBAN-1442-5GEA 0 GBP
COBBAN-1442-5G COBBAN-1442-1G
2-Bromo-4-nitroaniline
2-Bromo-4-nitroaniline
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD00025152
CAS Number: 13296-94-1
EINECS: 236-318-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR