2,6-Dibromo-4-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Amino-3,5-dibromo-α,α,α-trifluorotoluene
AN-1559-25G
AN-1559-1G
AN-1559-5G
COBBAN-1559-25GEA
0
GBP
COBBAN-1559-25G
COBBAN-1559-1G
COBBAN-1559-5G
2,6-Dibromo-4-(trifluoromethyl)aniline
2,6-Dibromo-4-(trifluoromethyl)aniline
Formula:
C₇H₄Br₂F₃N Boiling Pt: 64…65 °C (0.1 mmHg) Melting Pt: 38…40 °C |
MDL Number:
MFCD00068181 CAS Number: 72678-19-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...