2,5-Dibromoaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1522-5G
AN-1522-25G
COBBAN-1522-5GEA
0
GBP
COBBAN-1522-5G
COBBAN-1522-25G
2,5-Dibromoaniline
2,5-Dibromoaniline
Formula:
C₆H₅Br₂N MW: 250.92 g/mol Boiling Pt: 281 Melting Pt: 54.5…55 °C |
MDL Number:
MFCD00007636 CAS Number: 3638-73-1 EINECS: 222-865-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...