2,3-Difluoroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7615-1G
OS-7615-100G
OS-7615-25G
OS-7615-5G
COBBOS-7615-1GEA
0
GBP
COBBOS-7615-1G
COBBOS-7615-100G
COBBOS-7615-25G
COBBOS-7615-5G
2,3-Difluoroaniline
2,3-Difluoroaniline
Formula:
C₆H₅F₂N Boiling Pt: 69 °C (12 mmHg) Density: 1.274 g/cm³ (20 °C) |
MDL Number:
MFCD00010298 CAS Number: 4519-40-8 EINECS: 224-847-2 UN: 2941 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...