7-Isopropyl-1,4-dimethylazulene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
CS-9192-5G
CS-9192-25G
CS-9192-100G
COBBCS-9192-5GEA
0
GBP
COBBCS-9192-5G
COBBCS-9192-25G
COBBCS-9192-100G
7-Isopropyl-1,4-dimethylazulene
7-Isopropyl-1,4-dimethylazulene
Formula:
C₁₅H₁₈ Boiling Pt: 309 °C (1013 hPa) Melting Pt: 133 °C Density: 0.9759 g/cm³ (18 °C) |
MDL Number:
MFCD00003811 CAS Number: 489-84-9 EINECS: 207-701-2 Merck Index: 12,04581 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...