1,4-Dibromo-2-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AN-1087-5G AN-1087-25G
COBBAN-1087-5GEA 0 GBP
COBBAN-1087-5G COBBAN-1087-25G
1,4-Dibromo-2-nitrobenzene
1,4-Dibromo-2-nitrobenzene
Formula: C₆H₃Br₂NO₂
Melting Pt: 81…85 °C
Density: 2,374 g/cm³ (20 °C)
MDL Number: MFCD00007046
CAS Number: 3460-18-2
EINECS: 222-404-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR