Piperazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AM-1616-25G
AM-1616-100G
COBBAM-1616-25GEA
0
GBP
COBBAM-1616-25G
COBBAM-1616-100G
Piperazine
Piperazine
Formula:
C₄H₁₀N₂ MW: 86.14 g/mol Boiling Pt: 146 °C (1013 hPa) Melting Pt: 44…46 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005953 CAS Number: 110-85-0 EINECS: 203-808-3 UN: 2579 ADR: 8,III Merck Index: 13,07545 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...