2',6'-Difluoro-3'-methylacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(2,6-Difluoro-3-methylphenyl)ethanone
005743-1G
005743-5G
FLUO005743-1GEA
0
GBP
FLUO005743-1G
FLUO005743-5G
2',6'-Difluoro-3'-methylacetophenone
2',6'-Difluoro-3'-methylacetophenone
Formula:
C₉H₈F₂O |
MDL Number:
MFCD01631339 CAS Number: 261763-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...