2,3-Difluoroanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OT-0544-25G OT-0544-5G
COBBOT-0544-25GEA 0 GBP
COBBOT-0544-25G COBBOT-0544-5G
2,3-Difluoroanisole
2,3-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144.12 g/mol
MDL Number: MFCD01631562
CAS Number: 134364-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR