Tris(perfluorobutyl)amine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007167-25G 007167-100G
FLUO007167-25GEA 0 GBP
FLUO007167-25G FLUO007167-100G
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –55 °C
Density: 1,883 g/cm³ (20 °C)
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR