4-(Trifluoromethyl)phenylacetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007637-5G
007637-25G
007637-100G
FLUO007637-5GEA
0
GBP
FLUO007637-5G
FLUO007637-25G
FLUO007637-100G
4-(Trifluoromethyl)phenylacetonitrile
4-(Trifluoromethyl)phenylacetonitrile
Formula:
C₉H₆F₃N Boiling Pt: 131…132 °C (20 mmHg) Melting Pt: 47.85…48.85 °C Density: 1.535 g/cm³ (–150.15 °C) |
MDL Number:
MFCD00001921 CAS Number: 2338-75-2 EINECS: 219-052-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...