4-(Trifluoromethoxy)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007566-1G 007566-5G 007566-25G
FLUO007566-1GEA 0 GBP
FLUO007566-1G FLUO007566-5G FLUO007566-25G
4-(Trifluoromethoxy)phenylacetonitrile
4-(Trifluoromethoxy)phenylacetonitrile
Formula: C₉H₆F₃NO
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00061240
CAS Number: 49561-96-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR