2-(Trifluoromethoxy)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007984-1G 007984-5G
FLUO007984-1GEA 0 GBP
FLUO007984-1G FLUO007984-5G
2-(Trifluoromethoxy)phenylacetonitrile
2-(Trifluoromethoxy)phenylacetonitrile
Formula: C₉H₆F₃NO
MDL Number: MFCD00236326
CAS Number: 137218-25-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR