α-Bromo-2-iodotoulene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OS-7704-100G
OS-7704-5G
OS-7704-1G
OS-7704-25G
COBBOS-7704-100GEA
0
GBP
COBBOS-7704-100G
COBBOS-7704-5G
COBBOS-7704-1G
COBBOS-7704-25G
α-Bromo-2-iodotoulene
α-Bromo-2-iodotoulene
Formula:
C₇H₆BrI Boiling Pt: 279 °C (1013 hPa) Melting Pt: 55.5 °C |
MDL Number:
MFCD00236046 CAS Number: 40400-13-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...