1-(4-Nitrophenyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018909-1G 018909-5G
FLUO018909-1GEA 0 GBP
FLUO018909-1G FLUO018909-5G
1-(4-Nitrophenyl)imidazole
Formula: C₉H₇N₃O₂
MDL Number: MFCD00060491
CAS Number: 2301-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR