1-(4-Methoxyphenyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018799-1G 018799-5G
FLUO018799-1GEA 0 GBP
FLUO018799-1G FLUO018799-5G
1-(4-Methoxyphenyl)imidazole
1-(4-Methoxyphenyl)imidazole
Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00060492
CAS Number: 10040-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR