1-(2-Hydroxyethyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005948-5G 005948-25G
FLUO005948-5GEA 0 GBP
FLUO005948-5G FLUO005948-25G
1-(2-Hydroxyethyl)imidazole
1-(2-Hydroxyethyl)imidazole
Formula: C₅H₈N₂O
MDL Number: MFCD00209980
CAS Number: 1615-14-1
EINECS: 216-565-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR