Dopamine hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008896-5G 008896-25G
FLUO008896-5GEA 0 GBP
FLUO008896-5G FLUO008896-25G
Dopamine hydrochloride
Dopamine hydrochloride
Formula: C₈H₁₂ClNO₂
Melting Pt: 246…249 °C
Density: 1,356 g/cm³ (20 °C)
MDL Number: MFCD00012898
CAS Number: 62-31-7
EINECS: 200-527-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR