Bis(pentafluorophenyl)carbonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bis(pentafluorophenyl) Carbonate

010236-5G 010236-25G
FLUO010236-5GEA 0 GBP
FLUO010236-5G FLUO010236-25G
Bis(pentafluorophenyl)carbonate
Bis(pentafluorophenyl) carbonate
Formula: C₁₃F₁₀O₃
MW: 394.13 g/mol
MDL Number: MFCD00368353
CAS Number: 59483-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR