tert-Butyl carbamate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Carbamic acid tert-butyl ester

OS-7686-100G OS-7686-25G OS-7686-5G
COBBOS-7686-100GEA 0 GBP
COBBOS-7686-100G COBBOS-7686-25G COBBOS-7686-5G
tert-Butyl carbamate
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR