Bicine

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

9162.1 9162.2
ROTH9162.1EA 0 GBP
ROTH9162.1 ROTH9162.2
Bicine
Bicine
Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
Melting Pt: 185…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR