Tributylallyltin, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane

M-Clarity™ quality level = MQ100

271411-100G 271411-25G 271411-5G
SIAL271411-100GEA 580 GBP
SIAL271411-100G SIAL271411-25G SIAL271411-5G
Tributylallyltin, Sigma-Aldrich®
Tributylallyltin
Formula: C₁₅H₃₂Sn
Boiling Pt: 88 °C (1013 hPa)
Density: 1.073 g/cm³ (25 °C)
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR