Tributylallyltin ≥97%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane
Purity: 97%
CAS No.: 24850-33-7
Molecular Formula: C15H32Sn
Formula Weight: 331.12
Storage Temperature: 2°C to 8°C
Flash Point: 103°(217°F)
Refractive Index: 1.4860
Boiling Point: 100-102°/0.5mm
Beilstein Reference: 3588340
UN No.: 2788
MDL No.: MFCD00010346
Formula:
C₁₅H₃₂Sn Boiling Pt: 100…102 °C (0.5 mmHg) Density: 1.079 Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00010346 CAS Number: 24850-33-7 EINECS: 246-494-3 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...