Bromoacetaldehyde diethyl acetal
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-1,1-diethoxyethane
003024-100G
003024-500G
FLUO003024-100GEA
0
GBP
FLUO003024-100G
FLUO003024-500G
Bromoacetaldehyde diethyl acetal
Bromoacetaldehyde diethyl acetal
Formula:
BrH₂CCH(OCH₂CH₃)₂ MW: 197.07 g/mol Boiling Pt: 66 °C (1013 hPa) Density: 1.27 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000214 CAS Number: 2032-35-1 EINECS: 217-989-1 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...