3-Bromo-1,1,1-trifluoroacetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001268-10G 001268-25G 001268-100G
FLUO001268-10GEA 0 GBP
FLUO001268-10G FLUO001268-25G FLUO001268-100G
3-Bromo-1,1,1-trifluoroacetone
3-Bromo-1,1,1-trifluoroacetone
Formula: BrH₂CC(O)CF₃
Boiling Pt: 87 °C (1013 hPa)
Density: 1.8389 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR